Learn More
New England Biolabs, Inc. NEBNext® High-Fidelity 2X PCR Master Mix – 250 reactions
SDP
Catalog No. NC0373959
Encompass
NEBNext High-Fidelity 2X PCR Master Mix is the original non-hot start NEBNext formulation of Q5 High-Fidelity DNA Polymerase. This formulation is also included as a component in the original NEBNext standard workflow library prep kits for Illumina, and in NEBNext library prep kits for Ion Torrent.
  • Next generation sequencing library amplification
  • High fidelity PCR
  • High-throughput PCR
Catalog No. NC0373959 Supplier New England Biolabs, Inc. Supplier No. M0541L
NEBNext High-Fidelity 2X PCR Master Mix is the original non-hot start NEBNext formulation of Q5 High-Fidelity DNA Polymerase. This formulation is also included as a component in the original NEBNext standard workflow library prep kits for Illumina, and in NEBNext library prep kits for Ion Torrent.
  • Next generation sequencing library amplification
  • High fidelity PCR
  • High-throughput PCR