Learn More
New England Biolabs, Inc. NEBNext Poly(A) mRNA Magnetic Isolation Module – 96 reactions
SDP
Catalog No. NC0477294
Encompass
The NEBNext Poly(A) mRNA Magnetic Isolation Module is designed to isolate intact poly(A) RNA from previously isolated total RNA. The technology is based on the coupling of Oligo d(T)25 to 1µm paramagnetic beads which is then used as the solid support for the direct binding of poly(A) RNA. Thus, the procedure permits the manual processing of multiple samples and can be adapted for automated high-throughput applications. Additionally, magnetic separation technology permits elution of intact mRNA in small volumes eliminating the need for precipitating the poly(A) transcripts in the eluent. Intact poly(A) RNA which is fully representative of the mRNA population of the original sample can be obtained in less than one hour.
Catalog No. NC0477294 Supplier New England Biolabs, Inc. Supplier No. E7490L
The NEBNext Poly(A) mRNA Magnetic Isolation Module is designed to isolate intact poly(A) RNA from previously isolated total RNA. The technology is based on the coupling of Oligo d(T)25 to 1µm paramagnetic beads which is then used as the solid support for the direct binding of poly(A) RNA. Thus, the procedure permits the manual processing of multiple samples and can be adapted for automated high-throughput applications. Additionally, magnetic separation technology permits elution of intact mRNA in small volumes eliminating the need for precipitating the poly(A) transcripts in the eluent. Intact poly(A) RNA which is fully representative of the mRNA population of the original sample can be obtained in less than one hour.