Learn More
New England Biolabs, Inc. NEBNext Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1-48) – 96 reactions
SDP
Catalog No. NC1250970
Encompass
The NEBNext Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1-48) contains the adaptors, primers, enzymes and buffers required to convert small RNAs into indexed libraries for next-generation sequencing on the Illumina platform. The novel workflow has been optimized to minimize adaptor-dimers, while producing high-yield, high-diversity libraries.
Catalog No. NC1250970 Supplier New England Biolabs, Inc. Supplier No. E7560S
Add to Cart
Add to Cart
Request a Quote
The NEBNext Multiplex Small RNA Library Prep Kit for Illumina (Index Primers 1-48) contains the adaptors, primers, enzymes and buffers required to convert small RNAs into indexed libraries for next-generation sequencing on the Illumina platform. The novel workflow has been optimized to minimize adaptor-dimers, while producing high-yield, high-diversity libraries.