Learn More
New England Biolabs, Inc. NEB 10-beta/Stable Outgrowth Medium – 100 ml (4 x 25 ml)
SDP
Catalog No. 501526334
Encompass
Add NEB 10-beta/Stable Outgrowth Medium to freshly transformed NEB 10-beta or NEB Stable E. coli cells and incubate for up to 1 hour before plating on selective media. This medium can also be used to dilute the outgrowth prior to plating.
Catalog No. 50-152-6334 Supplier New England Biolabs, Inc. Supplier No. B9035S
Add NEB 10-beta/Stable Outgrowth Medium to freshly transformed NEB 10-beta or NEB Stable E. coli cells and incubate for up to 1 hour before plating on selective media. This medium can also be used to dilute the outgrowth prior to plating.