Learn More
TARGETMOL CHEMICALS INC Nebentan potassium 2MG
SDP
Catalog No. 502879336
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Nebentan potassium (YM598) is an orally active and selective nonpeptide endothelin receptor (ETA receptor) antagonist.Nebentan potassium inhibits pulmonary arterial hypertension and may be useful in the study of neurologic and cardiovascular diseases. Purity 98.92%
Catalog No. 50-287-9336 Supplier TARGETMOL CHEMICALS INC Supplier No. T360082MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Nebentan potassium (YM598) is an orally active and selective nonpeptide endothelin receptor (ETA receptor) antagonist.Nebentan potassium inhibits pulmonary arterial hypertension and may be useful in the study of neurologic and cardiovascular diseases. Purity 98.92%