Learn More
New England Biolabs, Inc. NEBExpress Cell-Free E. coli Protein Synthesis System – 100 reactions
SDP
Catalog No. 502024006
Encompass
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17-230 kDa in just 2-4 hours under the control of T7 RNA Polymerase.
Catalog No. 50-202-4006 Supplier New England Biolabs, Inc. Supplier No. E5360L
The NEBExpress Cell-free E. coli Protein Synthesis System is an extract-based transcription/translation system derived from E. coli cells that have been engineered for high in vitro synthesis performance. It is designed to synthesize proteins ranging in size from 17-230 kDa in just 2-4 hours under the control of T7 RNA Polymerase.