Learn More
New England Biolabs, Inc. NEBExpress Ni Resin – 25 ml
SDP
Catalog No. 501959409
Encompass
NEBExpress Ni Resin is an affinity matrix for the isolation and purification of polyhistidine-tagged (His-tagged) fusion proteins. It is intended for use in gravity or pressure flow columns, and batch purifications. NEBExpress Ni Resin is comprised of a highly uniform and stable chemical-tolerant resin, pre-charged with nickel ions on the matrix surface. It is resistant to a wide range of chemicals, including NaOH, EDTA, DTT and beta-Mercaptoethanol.
Catalog No. 50-195-9409 Supplier New England Biolabs, Inc. Supplier No. S1428S
NEBExpress Ni Resin is an affinity matrix for the isolation and purification of polyhistidine-tagged (His-tagged) fusion proteins. It is intended for use in gravity or pressure flow columns, and batch purifications. NEBExpress Ni Resin is comprised of a highly uniform and stable chemical-tolerant resin, pre-charged with nickel ions on the matrix surface. It is resistant to a wide range of chemicals, including NaOH, EDTA, DTT and beta-Mercaptoethanol.