Learn More
Medchemexpress LLC Nebivolol hydrochloride | 152520-56-4 | 99.9% | 1 ML
SDP
Catalog No. 5000406643
Encompass_Preferred
Nebivolol hydrochloride (R 065824) is an orally active beta receptor blocker with high beta(1)-receptor affinity. It possesses direct vasodilator properties and adrenergic blocking characteristics.
  • Used for research into various diseases
  • High beta(1)-receptor affinity
  • Direct vasodilator properties
  • Adrenergic blocking characteristics
  • Orally active beta receptor blocker
Catalog No. 50-004-06643 Supplier Medchemexpress LLC Supplier No. HYB0203A10MM/1ML
Nebivolol hydrochloride (R 065824) is an orally active beta receptor blocker with high beta(1)-receptor affinity. It possesses direct vasodilator properties and adrenergic blocking characteristics.
  • Used for research into various diseases
  • High beta(1)-receptor affinity
  • Direct vasodilator properties
  • Adrenergic blocking characteristics
  • Orally active beta receptor blocker