Learn More
Medchemexpress LLC Nebivolol hydrochloride | 152520-56-4 | MFCD07357273 | 99.9% | 50 MG
SDP
Catalog No. 5000407413
Encompass_Preferred
Nebivolol hydrochloride is an orally active beta receptor blocker with high beta(1)-receptor affinity. It possesses direct vasodilator properties and adrenergic blocking characteristics. This compound is used for the research of various diseases such as hypertension, coronary artery disease, congestive heart failure, and ischemic heart disease.
  • Orally active beta receptor blocker
  • High beta(1)-receptor affinity
  • Direct vasodilator properties
  • Adrenergic blocking characteristics
  • Improves endothelial dysfunction
  • Stimulates endothelial nitric oxide synthase activity
  • Antioxidative properties
  • Inhibits haCSMC and haEC proliferation
  • Induces moderate apoptosis
  • Increases NO formation and decreases endothelin-1 secretion
Catalog No. 50-004-07413 Supplier Medchemexpress LLC Supplier No. HYB0203A50MG
Nebivolol hydrochloride is an orally active beta receptor blocker with high beta(1)-receptor affinity. It possesses direct vasodilator properties and adrenergic blocking characteristics. This compound is used for the research of various diseases such as hypertension, coronary artery disease, congestive heart failure, and ischemic heart disease.
  • Orally active beta receptor blocker
  • High beta(1)-receptor affinity
  • Direct vasodilator properties
  • Adrenergic blocking characteristics
  • Improves endothelial dysfunction
  • Stimulates endothelial nitric oxide synthase activity
  • Antioxidative properties
  • Inhibits haCSMC and haEC proliferation
  • Induces moderate apoptosis
  • Increases NO formation and decreases endothelin-1 secretion