Learn More
Selleck Chemical LLC Nebivolol hydrochloride (R-65824) S1549-10mM/1mL
SDP
Catalog No. 503108651
Encompass_Preferred
Nebivolol HCl (R-65824) selectively inhibits 1-adrenoceptor with IC50 of 0 8 nM
Catalog No. 50-310-8651 Supplier Selleck Chemical LLC Supplier No. S154910MM/1ML
Nebivolol HCl (R-65824) selectively inhibits 1-adrenoceptor with IC50 of 0 8 nM