Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Nebivolol hydrochloride

Catalog No. AC462492500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462492500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462492500 Supplier Thermo Scientific Chemicals Supplier No. 462492500

missing translation for 'validMainframeMsg'

CAS: 152520-56-4 | C22H26ClF2NO4 | 441.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152520-56-4
Molecular Formula C22H26ClF2NO4
Molecular Weight (g/mol) 441.90
InChI Key JWEXHQAEWHKGCW-BIISKSHESA-N
IUPAC Name hydrogen (1S)-1-[(2S)-6-fluoro-3,4-dihydro-2H-1-benzopyran-2-yl]-2-{[(2S)-2-[(2R)-6-fluoro-3,4-dihydro-2H-1-benzopyran-2-yl]-2-hydroxyethyl]amino}ethan-1-ol chloride
SMILES [H+].[Cl-].O[C@@H](CNC[C@H](O)[C@@H]1CCC2=CC(F)=CC=C2O1)[C@H]1CCC2=CC(F)=CC=C2O1

Specifications

Melting Point 226.0°C
Quantity 250 mg
Formula Weight 441.9
Chemical Name or Material Nebivolol hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.