Learn More
New England Biolabs, Inc. NEBNext Cell Lysis Buffer Module – 96 reactions
SDP
Catalog No. 501959352
Encompass
This module contains the enzymes and buffers required to lyse isolated cultured and primary cells to extract RNA. The module is optimized for use with the NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina or NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, and can be used to supplement the NEBNext Cell Lysis Buffer and Murine RNase Inhibitor components supplied in these products.
Catalog No. 50-195-9352 Supplier New England Biolabs, Inc. Supplier No. E5530S
This module contains the enzymes and buffers required to lyse isolated cultured and primary cells to extract RNA. The module is optimized for use with the NEBNext Single Cell/Low Input RNA Library Prep Kit for Illumina or NEBNext Single Cell/Low Input cDNA Synthesis and Amplification Module, and can be used to supplement the NEBNext Cell Lysis Buffer and Murine RNase Inhibitor components supplied in these products.