Learn More
New England Biolabs, Inc. NEBNext ARTIC SARS-CoV-2 Compantion Kit (Oxford Nanopore Technologies) - 96 rxns
SDP
Supplier:  New England Biolabs, Inc. E7660L
Encompass
The NEBNext ARTIC kits were developed in response to the critical need for reliable and accurate methods for sequencing SARS-CoV-2. These kits, for long and short read sequencing, were based on the original work of the ARTIC Network. The ARTIC SARS-CoV-2 sequencing workflow is a multiplexed amplicon-based whole-viral-genome sequencing approach. NEBNext ARTIC kits include primers and reagents for RT-PCR from SARS-CoV-2 gRNA and downstream library preparation. The balanced primers and reagents for RT-PCR deliver improved uniformity of amplicon yields from gRNA across a wide copy number range. The NEBNext ARTIC SARS-CoV-2 Companion Library Prep Kit (Oxford Nanopore Technologies) is compatible with a broad range of total RNA input amounts and is used alongside kits from Oxford Nanopore Technologies to generate targeted, high quality libraries for next-generation sequencing on the Oxford Nanopore platform.
Catalog No. 50-210-8557