Learn More
New England Biolabs, Inc. NEBNext dsDNA Fragmentase – 50 reactions
SDP
Catalog No. NC1001627
Encompass
NEBNext dsDNA Fragmentase is an enzyme-based reagent that shears DNA to produce fragments of the desired sizes in a time-dependent manner, for next generation sequencing library preparation protocols Note: NEBNext dsDNA Fragmentase is not part of the NEBNext Ultra II FS (Fragmentation System) reagents.
Catalog No. NC1001627 Supplier New England Biolabs, Inc. Supplier No. M0348S
NEBNext dsDNA Fragmentase is an enzyme-based reagent that shears DNA to produce fragments of the desired sizes in a time-dependent manner, for next generation sequencing library preparation protocols Note: NEBNext dsDNA Fragmentase is not part of the NEBNext Ultra II FS (Fragmentation System) reagents.