Learn More
New England Biolabs, Inc. NEBNext FFPE DNA Repair Mix – 24 reactions
SDP
Catalog No. 501047505
Encompass
The NEBNext FFPE DNA Repair Mix contains enzymes and buffers that are optimized to repair FFPE DNA in next generation sequencing workflows. The mix can be used upstream of any next generation sequencing library construction method, for any sequencing platform.
Catalog No. 50-104-7505 Supplier New England Biolabs, Inc. Supplier No. M6630S
The NEBNext FFPE DNA Repair Mix contains enzymes and buffers that are optimized to repair FFPE DNA in next generation sequencing workflows. The mix can be used upstream of any next generation sequencing library construction method, for any sequencing platform.