Learn More
New England Biolabs, Inc. NEBNext FFPE Library Prep 24
SDP
Catalog No. 502527898
Encompass
The NEBNext FFPE DNA Library Prep Kit includes the NEBNext FFPE DNA Repair v2 Mix, an optimized cocktail of enzymes designed to repair FFPE DNA, library prep reagents featuring a new polymerase master mix, and a protocol optimized for FFPE DNA. This kit is designed for use with pre-sheared FFPE DNA.
Catalog No. 50-252-7898 Supplier New England Biolabs, Inc. Supplier No. E6650S
The NEBNext FFPE DNA Library Prep Kit includes the NEBNext FFPE DNA Repair v2 Mix, an optimized cocktail of enzymes designed to repair FFPE DNA, library prep reagents featuring a new polymerase master mix, and a protocol optimized for FFPE DNA. This kit is designed for use with pre-sheared FFPE DNA.