Learn More
New England Biolabs, Inc. NEBNext FFPE DNA Repair v2 Module 96 rxns
SDP
Catalog No. 502253073
Encompass
This new product further improves repair of FFPE DNA in next generation sequencing (NGS) workflows. The methods used for fixation and storage of Formalin-Fixed Paraffin-Embedded (FFPE) DNA samples cause significant damage making it challenging to obtain high quality sequence data. The NEBNext FFPE DNA Repair v2 Module is an optimized cocktail of enzymes designed to repair FFPE DNA and supplied with optimized reagents to enable a streamlined workflow for NGS library preparation.
This item is not returnable. View return policy
Catalog No. 50-225-3073 Supplier New England Biolabs, Inc. Supplier No. E7360L
This new product further improves repair of FFPE DNA in next generation sequencing (NGS) workflows. The methods used for fixation and storage of Formalin-Fixed Paraffin-Embedded (FFPE) DNA samples cause significant damage making it challenging to obtain high quality sequence data. The NEBNext FFPE DNA Repair v2 Module is an optimized cocktail of enzymes designed to repair FFPE DNA and supplied with optimized reagents to enable a streamlined workflow for NGS library preparation.