Learn More
New England Biolabs, Inc. NEBNext High Input Poly(A) mRNA Isolation Module - 24 reactions
SDP
Catalog No. 502336250
Encompass
The NEBNext High-Input Poly(A) mRNA Magnetic Isolation Module is designed to isolate intact poly(A)+ RNA from high inputs (5-50ug) of previously isolated total RNA. The technology is based on the coupling of oligo d(T)25 to paramagnetic beads which is then used as the solid support for the direct binding of poly(A)+ RNA. The procedure permits the manual processing of multiple samples and can be adapted for automated high-throughput applications. Additionally, magnetic separation technology permits elution of intact mRNA in small volumes eliminating the need for precipitating the poly(A)+ transcripts in the eluent. Intact poly(A)+ RNA which is representative of the mRNA population of the original sample can be obtained in approximately one hour.
Catalog No. 50-233-6250 Supplier New England Biolabs, Inc. Supplier No. E3370S
The NEBNext High-Input Poly(A) mRNA Magnetic Isolation Module is designed to isolate intact poly(A)+ RNA from high inputs (5-50ug) of previously isolated total RNA. The technology is based on the coupling of oligo d(T)25 to paramagnetic beads which is then used as the solid support for the direct binding of poly(A)+ RNA. The procedure permits the manual processing of multiple samples and can be adapted for automated high-throughput applications. Additionally, magnetic separation technology permits elution of intact mRNA in small volumes eliminating the need for precipitating the poly(A)+ transcripts in the eluent. Intact poly(A)+ RNA which is representative of the mRNA population of the original sample can be obtained in approximately one hour.