Learn More
New England Biolabs, Inc. NEBNext Library Dilution Buffer – 7.5 ml
SDP
Catalog No. 501047524
Encompass
The NEBNext Library Dilution Buffer is optimized for dilution of the libraries in the workflow for the NEBNext Library Quant Kit.
Catalog No. 50-104-7524 Supplier New England Biolabs, Inc. Supplier No. B6118S
The NEBNext Library Dilution Buffer is optimized for dilution of the libraries in the workflow for the NEBNext Library Quant Kit.