Learn More
New England Biolabs, Inc. NEBNext Low-bias Small RNA Library Prep Kit - 24 reactions
SDP
Catalog No. 5000077835
Encompass
NEBNext Low-bias Small RNA Library Prep Kit reliably generates high-quality NGS libraries from total (0.5 ng - 1,000 ng) or enriched small (0.05 ng - 5 ng) RNA. The kit's reagents and workflow together minimize the amount of bias exhibited by the resulting libriaries. This means that the libraries are more accurately reflective of the various small RNA species and their relative proportions in the sample. For optimal flexibility in multiplexing, the NEBNext Low-bias Small RNA Library Prep Kit is designed to work with the NEBNext LV Unique Dual Index Primers (Sets 1-5), which must be purchased separately; 480 indices are available.
  • Generates libraries with the lowest bias among commercially available kits
  • Fastest workflow (~3.5 hours) for single-day libraries
  • Multiplexable with up to 480 UDIs
  • Simplified, gel-free workflow
  • 18-month shelf life
Catalog No. 50-000-77835 Supplier New England Biolabs, Inc. Supplier No. E3420S
NEBNext Low-bias Small RNA Library Prep Kit reliably generates high-quality NGS libraries from total (0.5 ng - 1,000 ng) or enriched small (0.05 ng - 5 ng) RNA. The kit's reagents and workflow together minimize the amount of bias exhibited by the resulting libriaries. This means that the libraries are more accurately reflective of the various small RNA species and their relative proportions in the sample. For optimal flexibility in multiplexing, the NEBNext Low-bias Small RNA Library Prep Kit is designed to work with the NEBNext LV Unique Dual Index Primers (Sets 1-5), which must be purchased separately; 480 indices are available.
  • Generates libraries with the lowest bias among commercially available kits
  • Fastest workflow (~3.5 hours) for single-day libraries
  • Multiplexable with up to 480 UDIs
  • Simplified, gel-free workflow
  • 18-month shelf life