Learn More
New England Biolabs, Inc. NEBNext Magnetic Separation Rack – 24 tubes
SDP
Catalog No. 501959387
Encompass
Next generation sequencing library preparation workflows include magnetic bead-based purification and size-selection steps. It is important for library yield and quality that bead separation be highly efficient and fast. This rack was designed for this application and contains rare earth Neodymium Iron Boron (NdFeB) magnets, the most powerful commercially available magnets, in an anodized aluminum rack. The rack holds 24 0.2ml tubes, and is compatible with single tubes or strip tubes.
Catalog No. 50-195-9387 Supplier New England Biolabs, Inc. Supplier No. S1515S
Add to Cart
Add to Cart
Next generation sequencing library preparation workflows include magnetic bead-based purification and size-selection steps. It is important for library yield and quality that bead separation be highly efficient and fast. This rack was designed for this application and contains rare earth Neodymium Iron Boron (NdFeB) magnets, the most powerful commercially available magnets, in an anodized aluminum rack. The rack holds 24 0.2ml tubes, and is compatible with single tubes or strip tubes.