Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II Non-Directional RNA Second Strand Synthesis Module – 100 reactions
SDP
Catalog No. 504332
Encompass
This module is part of the non-directional Ultra II RNA workflow, for Illumina- compatible non-strand-specific RNA library construction. This workflow is compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enables high yield preparation of high quality libraries from 5 ng – 1 µg total RNA.
  • Designed for use in the NEBNext Ultra II non-directional RNA workflow
Catalog No. 50-433-2 Supplier New England Biolabs, Inc. Supplier No. E6111L
This module is part of the non-directional Ultra II RNA workflow, for Illumina- compatible non-strand-specific RNA library construction. This workflow is compatible with poly(A) mRNA isolation or ribosomal RNA depletion, and enables high yield preparation of high quality libraries from 5 ng – 1 µg total RNA.
  • Designed for use in the NEBNext Ultra II non-directional RNA workflow