Learn More
New England Biolabs, Inc. NEBNext Multiplex Oligos for Illumina (Index Primers Set 4) – 96 reactions
SDP
Catalog No. 501526370
Encompass
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.
Catalog No. 50-152-6370 Supplier New England Biolabs, Inc. Supplier No. E7730L
NEBNext Multiplex Oligos provide adaptors and primers to enable high yield multiplex Illumina library production. The unique hairpin loop structure of the NEBNext Adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.