Learn More
New England Biolabs, Inc. NEBNext Primers for Epigenetics (Unique Dual Index Set 2B) 24 rxns
SDP
Catalog No. 502551934
Encompass
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries. This 24-reaction kit is supplied in a plate with premixed i5 and i7 8-base index primers in 24 wells.
Catalog No. 50-255-1934 Supplier New England Biolabs, Inc. Supplier No. E3392S
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries. This 24-reaction kit is supplied in a plate with premixed i5 and i7 8-base index primers in 24 wells.