Learn More
New England Biolabs, Inc. NEBNext Primers for Epigenetics (Unique Dual Index Set 3) 96 rxns
SDP
Catalog No. 502551933
Encompass
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries. This 96 reaction kit contains premixed i5 and i7 8-base index primers, and each well of a 96-well plate contains an i5 and i7 primer pair.
Catalog No. 50-255-1933 Supplier New England Biolabs, Inc. Supplier No. E3404S
The NEBNext Primers for Epigenetics contain unique dual index primers optimized for E5hmC-seq libraries. This 96 reaction kit contains premixed i5 and i7 8-base index primers, and each well of a 96-well plate contains an i5 and i7 primer pair.