Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit (Human/Mouse/Rat) – 24 reactions
SDP
Supplier:  New England Biolabs, Inc. E6310L
Encompass
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human, mouse and rat total RNA preparations. This product is suitable for both intact and degraded RNA (e.g. FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications.
Catalog No. NC0863026
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product