Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit v2 (Human, Mouse, Rat) with RNA Sample Purification Beads – 6 reactions
SDP
Supplier:  New England Biolabs, Inc. E7405S
Encompass
The NEBNext rRNA Depletion Kit (Human/Mouse/Rat) with RNA Sample Purification Beads employs an RNaseH-based method to deplete both cytoplasmic (5S rRNA, 5.8S rRNA, 18S rRNA and 28S rRNA) and mitochondrial ribosomal RNA (12S rRNA and 16S rRNA) from human, mouse and rat total RNA preparations. This product is suitable for both intact and degraded RNA (e.g., FFPE RNA). The resulting rRNA-depleted RNA is suitable for RNA-Seq, random-primed cDNA synthesis, or other downstream RNA analysis applications. For added convenience, this kit contains NEBNext RNA Sample Purification Beads (Agencourt RNAClean XP beads from Beckman Coulter).
Catalog No. 50-202-4025