Learn More
New England Biolabs, Inc. NEBNext Second Strand Synthesis (dNTP-free) Reaction Buffer – 0.4 ml
SDP
Catalog No. 504287
Encompass
A 10X reaction buffer for use with NEBNext Second Strand Synthesis Enzyme Mix when alternative concentrations of deoxynucleotides are required
Catalog No. 50-428-7 Supplier New England Biolabs, Inc. Supplier No. B6117S
A 10X reaction buffer for use with NEBNext Second Strand Synthesis Enzyme Mix when alternative concentrations of deoxynucleotides are required