Learn More
New England Biolabs, Inc. NEBNext Ultra II FS DNA PCR-free Library Prep Kit for Illumina - 96 rxn
SDP
Catalog No. 502172591
Encompass
The NEBNext Ultra II DNA PCR-free Library Prep Kit contains the enzymes and buffers required to convert a broad range of intact DNA input amounts into libraries for next-generation sequencing on the Illumina platform all without an amplification step. The fast user-friendly workflow includes enzymatic fragmentation for optimal input size requires minimal hands-on time and maintains excellent library yields and GC coverage without requiring PCR. Dont be biased The NEBNext Ultra II FS DNA PCR-free Library Prep Kit for Illumina can save you time and minimize amplification-induced sequence bias and errors with a complete solution for PCR-free NGS library preparation. Built on the robust and reliable Ultra II reagents and workflow from New England Biolabs you can trust that your samples are in the best hands your own.
Catalog No. 50-217-2591 Supplier New England Biolabs, Inc. Supplier No. E7430L
The NEBNext Ultra II DNA PCR-free Library Prep Kit contains the enzymes and buffers required to convert a broad range of intact DNA input amounts into libraries for next-generation sequencing on the Illumina platform all without an amplification step. The fast user-friendly workflow includes enzymatic fragmentation for optimal input size requires minimal hands-on time and maintains excellent library yields and GC coverage without requiring PCR. Dont be biased The NEBNext Ultra II FS DNA PCR-free Library Prep Kit for Illumina can save you time and minimize amplification-induced sequence bias and errors with a complete solution for PCR-free NGS library preparation. Built on the robust and reliable Ultra II reagents and workflow from New England Biolabs you can trust that your samples are in the best hands your own.