Learn More
New England Biolabs, Inc. NEBNext® Ultra™ II Directional RNA Library Prep Kit for Illumina® – 24 reactions
SDP
Catalog No. NC1398112
Encompass
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the “dUTP” method for strand-specificity, with proven superiority for this application.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE
Catalog No. NC1398112 Supplier New England Biolabs, Inc. Supplier No. E7760S
The Ultra II Directional RNA Library Prep Kit for Illumina delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA. In conjunction with ribosomal RNA (rRNA) depletion or poly(A) enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. The NEBNext Ultra II Directional RNA Library Prep Kit derives its directionality from the “dUTP” method for strand-specificity, with proven superiority for this application.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE