Learn More
New England Biolabs, Inc. NEBNext Ultra™ II DNA Library Prep with Sample Purification Beads – 24 reactions
SDP
Catalog No. 501526362
Encompass
The NEBNext Ultra II DNA Library Prep with Sample Purification Beads contains the enzymes, buffers and purification beads required to convert a broad range of DNA input amounts into indexed libraries for next-generation sequencing on the Illumina platform. The fast, user-friendly workflow has minimal hands-on time and, for added convenience, this kit contains NEBNext Sample Purification Beads (SPRIselect beads from Beckman Coulter) for size selection and enzyme reaction cleanup.
Catalog No. 50-152-6362 Supplier New England Biolabs, Inc. Supplier No. E7103S
The NEBNext Ultra II DNA Library Prep with Sample Purification Beads contains the enzymes, buffers and purification beads required to convert a broad range of DNA input amounts into indexed libraries for next-generation sequencing on the Illumina platform. The fast, user-friendly workflow has minimal hands-on time and, for added convenience, this kit contains NEBNext Sample Purification Beads (SPRIselect beads from Beckman Coulter) for size selection and enzyme reaction cleanup.