Learn More
New England Biolabs, Inc. NEBNext UltraExpress DNA 24
SDP
Catalog No. 502527902
Encompass
The NEBNext UltraExpress DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.
Catalog No. 50-252-7902 Supplier New England Biolabs, Inc. Supplier No. E3325S
The NEBNext UltraExpress DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.