Learn More
New England Biolabs, Inc. NEBNext UltraExpress™ FS DNA Library Prep Kit - 96 rxns
SDP
Catalog No. 502551940
Encompass
The NEBNext UltraExpress FS DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow, including a fragmentation step. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range of intact DNA, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.
Catalog No. 50-255-1940 Supplier New England Biolabs, Inc. Supplier No. E3340L
The NEBNext UltraExpress FS DNA Library Prep Kit is the latest generation of NEBNext DNA library prep, with a fast, streamlined workflow, including a fragmentation step. In under 2 hours, the protocol enables the creation of high yield, high quality libraries, from a broad input amount range of intact DNA, while generating less plastic waste. Further, with simplicity at the forefront, the kit features a single protocol for all input amounts, making NEBNext UltraExpress DNA Library Prep well suited for all throughputs.