Learn More
New England Biolabs, Inc. NEBNext UltraExpress™ RNA Library Prep Kit 24 rxns
SDP
Catalog No. 502527900
Encompass
The NEBNext UltraExpress RNA Library Prep Kit is the latest generation of NEBNext RNA library prep, and has a fast, streamlined workflow. With a 3-hour library prep protocol, the kit enables creation of high-quality RNA libraries in a single day, in conjunction with mRNA enrichment or rRNA depletion kits.  
  • Fast workflow (3 hours)
  • Fewer steps and consumables
  • Fewer cleanups
  • Single protocol for all input amounts High-quality libraries from a broad input range: 25-250 ng total RNA
Catalog No. 50-252-7900 Supplier New England Biolabs, Inc. Supplier No. E3330S
The NEBNext UltraExpress RNA Library Prep Kit is the latest generation of NEBNext RNA library prep, and has a fast, streamlined workflow. With a 3-hour library prep protocol, the kit enables creation of high-quality RNA libraries in a single day, in conjunction with mRNA enrichment or rRNA depletion kits.  
  • Fast workflow (3 hours)
  • Fewer steps and consumables
  • Fewer cleanups
  • Single protocol for all input amounts High-quality libraries from a broad input range: 25-250 ng total RNA