Learn More
Medchemexpress LLC HY-113970A 5mg Medchemexpress, Nebracetam (hydrochloride) CAS:1177279-49-0 Purity:>98%
SDP
Catalog No. 501969275
Encompass_Preferred
Medchemexpress, HY-113970A 5mg Nebracetam (HCl) CAS:1177279-49-0 Nebracetam hydrochloride, a nootropic M1-muscarinic agonist, induces a rise of intracellular Ca2+ concentration. Nebracetam hydrochloride exhibits an EC50 of 1.59 mM for elevating [Ca2+]i. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9275 Supplier Medchemexpress LLC Supplier No. HY113970A5MG
Medchemexpress, HY-113970A 5mg Nebracetam (HCl) CAS:1177279-49-0 Nebracetam hydrochloride, a nootropic M1-muscarinic agonist, induces a rise of intracellular Ca2+ concentration. Nebracetam hydrochloride exhibits an EC50 of 1.59 mM for elevating [Ca2+]i. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.