Learn More
Medchemexpress LLC Nebracetam hydrochloride | 1177279-49-0 | 99.0% | 240.73 | 1 ML
SDP
Catalog No. 5000436819
Encompass_Preferred
Nebracetam hydrochloride (WEB 1881 FU) is an orally active M1 muscarinic receptor agonist that can increase intracellular Ca2+ concentration with an EC50 value of 1.59 mM. It demonstrates neuroprotective activity and can improve cognitive impairment, making it suitable for research into neurological diseases such as Alzheimer's disease.
  • Orally active M1 muscarinic receptor agonist.
  • Increases intracellular Ca2+ concentration with an EC50 value of 1.59 mM.
  • Demonstrates neuroprotective activity.
  • Improves cognitive impairment.
  • Used in research for neurological diseases such as Alzheimer's disease.
Catalog No. 50-004-36819 Supplier Medchemexpress LLC Supplier No. HY113970A10MM/1ML
Nebracetam hydrochloride (WEB 1881 FU) is an orally active M1 muscarinic receptor agonist that can increase intracellular Ca2+ concentration with an EC50 value of 1.59 mM. It demonstrates neuroprotective activity and can improve cognitive impairment, making it suitable for research into neurological diseases such as Alzheimer's disease.
  • Orally active M1 muscarinic receptor agonist.
  • Increases intracellular Ca2+ concentration with an EC50 value of 1.59 mM.
  • Demonstrates neuroprotective activity.
  • Improves cognitive impairment.
  • Used in research for neurological diseases such as Alzheimer's disease.