Learn More
Medchemexpress LLC Nebracetam hydrochloride | 1177279-49-0 | 99.0% | 240.73 | 10 MG
SDP
Catalog No. 5000436557
Encompass_Preferred
Nebracetam hydrochloride is an orally active M1 muscarinic receptor agonist. It induces an increase in intracellular Ca2+ concentration, with an EC50 value of 1.59 mM. This compound exhibits neuroprotective activity and can improve cognitive impairment, making it suitable for research into neurological diseases such as Alzheimer's disease.
  • Orally active M1 muscarinic receptor agonist
  • Induces an increase in intracellular Ca2+ concentration
  • Exhibits neuroprotective activity
  • Improves cognitive impairment
  • Suitable for research into neurological diseases
Catalog No. 50-004-36557 Supplier Medchemexpress LLC Supplier No. HY113970A10MG
Nebracetam hydrochloride is an orally active M1 muscarinic receptor agonist. It induces an increase in intracellular Ca2+ concentration, with an EC50 value of 1.59 mM. This compound exhibits neuroprotective activity and can improve cognitive impairment, making it suitable for research into neurological diseases such as Alzheimer's disease.
  • Orally active M1 muscarinic receptor agonist
  • Induces an increase in intracellular Ca2+ concentration
  • Exhibits neuroprotective activity
  • Improves cognitive impairment
  • Suitable for research into neurological diseases