Learn More
New England Biolabs, Inc. NEBridge Ligase MM - 250 rxn
SDP
Catalog No. 502527886
Encompass
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes, this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer. Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions, as well as moderate complexity (3-6 fragment) and high complexity (7-25 fragment) assemblies, are all supported with this optimized reagent and accompanying protocols.
Catalog No. 50-252-7886 Supplier New England Biolabs, Inc. Supplier No. M1100L
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes, this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer. Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions, as well as moderate complexity (3-6 fragment) and high complexity (7-25 fragment) assemblies, are all supported with this optimized reagent and accompanying protocols.