Learn More
New England Biolabs, Inc. NEBridge Ligase Master Mix - 50 rxn
SDP
Catalog No. 502172601
Encompass
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer. Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions as well as moderate complexity (3-6 fragment) and high complexity (7-25 fragment) assemblies are all supported with this optimized reagent and accompanying protocols.
Catalog No. 50-217-2601 Supplier New England Biolabs, Inc. Supplier No. M1100S
NEBridge Ligase Master Mix is a 3X master mix for Golden Gate Assembly. Designed for use with NEB Type IIS restriction enzymes this master mix contains T4 DNA Ligase in an optimized reaction buffer with a proprietary ligation enhancer. Users need only choose their preferred NEB Type IIS restriction enzyme and add DNA substrates to be assembled. Low complexity single fragment insertions as well as moderate complexity (3-6 fragment) and high complexity (7-25 fragment) assemblies are all supported with this optimized reagent and accompanying protocols.