Learn More
New England Biolabs, Inc. NEBuffer™ 1 – 5 ml
SDP
Catalog No. 50811549
Encompass
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. This NEBuffer minus BSA formulation is available for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation (which is included with NEB restriction enzymes), select NEBuffer 1.1. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.
Catalog No. 50-811-549 Supplier New England Biolabs, Inc. Supplier No. B7001S
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. This NEBuffer minus BSA formulation is available for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation (which is included with NEB restriction enzymes), select NEBuffer 1.1. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.