Learn More
New England Biolabs, Inc. NEBuffer™ 2 – 5 ml
SDP
Catalog No. 50811550
Encompass
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. This NEBuffer minus BSA formulation is available for those enzymes requiring NEBuffers without BSA. Note that BSA is no longer supplied with this product. For the BSA containing formulation (which is included with NEB restriction enzymes), select NEBuffer 2.1. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonucleases to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.
Catalog No. 50-811-550 Supplier New England Biolabs, Inc. Supplier No. B7002S
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. This NEBuffer minus BSA formulation is available for those enzymes requiring NEBuffers without BSA. Note that BSA is no longer supplied with this product. For the BSA containing formulation (which is included with NEB restriction enzymes), select NEBuffer 2.1. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonucleases to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1, NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.