Learn More
New England Biolabs, Inc. NEBuffer™ 3 – 5 ml
SDP
Catalog No. 50811551
Encompass
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1 , NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.
Catalog No. 50-811-551 Supplier New England Biolabs, Inc. Supplier No. B7003S
New England Biolabs provides a color-coded 10X NEBuffer (with BSA) with each restriction endonuclease to ensure optimal (100%) activity. New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. The NEBuffer minus BSA formulation is included in this buffer pack for those enzymes requiring NEBuffers without BSA. For the BSA containing formulation, Buffer packs NEBuffer 1.1 (NEB #B7201), NEBuffer 2.1 , NEBuffer 3.1 and CutSmart Buffer (NEB #B7204) are available.