Learn More
New England Biolabs, Inc. NEBuffer r2.1 - 5 mls
SDP
Catalog No. 502253070
Encompass
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB B9200).
This item is not returnable. View return policy
Catalog No. 50-225-3070 Supplier New England Biolabs, Inc. Supplier No. B6002S
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB B9200).