Learn More
New England Biolabs, Inc. NEBufferr3.1 - 5 mls
SDP
Catalog No. 502253071
Encompass
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB B9200).
This item is not returnable. View return policy
Catalog No. 50-225-3071 Supplier New England Biolabs, Inc. Supplier No. B6003S
New England Biolabs provides a color-coded 10X NEBuffer with each restriction endonuclease to ensure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. These buffers contain Recombinant Albumin (NEB B9200).