Learn More
New England Biolabs, Inc. NEBuffer™ Set (r1.1, r2.1, r3.1, rCutSmart™) 6 mls
SDP
Catalog No. 502108565
Encompass
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. This set contains NEBuffers that contain recombinant Albumin.
Catalog No. 50-210-8565 Supplier New England Biolabs, Inc. Supplier No. B7030S
New England Biolabs provides 10X NEBuffer with each restriction endonuclease to assure optimal (100%) activity. Most of our enzymes are supplied with one of four standard NEBuffers. This set contains NEBuffers that contain recombinant Albumin.