Learn More
Waters Corp Waters Corporation Nebulizer, ACQUITY UPLC ELSD
Catalog No. 50466536 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation Nebulizer is a critical component designed for the ACQUITY UPLC ELS Detector. It features a front-mounted, snap-in design and is temperature-controlled, essential for precise and reproducible measurements in UPLC applications.
  • Front-mounted design for easy installation.
  • Snap-in component for secure fitting.
  • Temperature-controlled for optimal performance.
  • Ensures precise measurements for UPLC applications.
  • Provides reproducible results for UPLC applications.
  • Categorized as a spare parts kit.
Catalog No. 50-466-536 Supplier Waters Corp Supplier No. 205000342
The Waters Corporation Nebulizer is a critical component designed for the ACQUITY UPLC ELS Detector. It features a front-mounted, snap-in design and is temperature-controlled, essential for precise and reproducible measurements in UPLC applications.
  • Front-mounted design for easy installation.
  • Snap-in component for secure fitting.
  • Temperature-controlled for optimal performance.
  • Ensures precise measurements for UPLC applications.
  • Provides reproducible results for UPLC applications.
  • Categorized as a spare parts kit.