Learn More
Precision Glassblowing of Colorado NEBULIZER AEROSALT HIGHSOLIDS
SDP
Catalog No. 50420747
Encompass_Preferred
Aerosalt High-Solids Nebulizer, 0.7 lpm Ar, 30 psi, 2 mL/min, Quick Disconnect, Argon Connect
Catalog No. 50-420-747 Supplier Precision Glassblowing of Colorado Supplier No. PG50065QDAC
Aerosalt High-Solids Nebulizer, 0.7 lpm Ar, 30 psi, 2 mL/min, Quick Disconnect, Argon Connect