Learn More
Med Vet International Nebulizer Compressor M08, J0949MN, Each
Catalog No. 5000583934
Encompass_Preferred
Nebulizer compressor unit with complete accessories. Uses room air to power a medicated nebulizer. Takes about 10 minutes. Mask not included. Uses 110V
Catalog No. 50-005-83934 Supplier Med Vet International Supplier No. JOR949M
Nebulizer compressor unit with complete accessories. Uses room air to power a medicated nebulizer. Takes about 10 minutes. Mask not included. Uses 110V