Learn More
Perkin Elmer US LLC Polyethylene Nebulizer Capillary Tubing, 0.58 I.D. - Qty. 10 ft
Supplier:  Perkin Elmer US LLC 09908265
Encompass_Preferred
This is a 10 foot (3 m) capillary tubing accessory for various nebulizers. Included in: Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer for Optima 8x00 (N0780607), Quick-Change Torch Module with Cyclonic Spray Chamber and Concentric Nebulizer for Optima 2x00/4x00/5x00/7x00 (N0770607), HF Sample Introduction Kit for NexION 300/350 (N8140507), Standard Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160143), Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle (N3160158), Universal GemTip Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370394), and the GemTip High-Sensitivity Plastic Body Nebulizer Complete Assembly for AAnalyst (N0370393). Note: This capillary tubing is not compatible with the High Sensitivity Nebulizer Assembly for AAnalyst 200/400/700/800 (N3150188).
Catalog No. NC9350400