Learn More
Glass Expansion Nebulizer gas fitting with ratchet connector for NexION 1000/2000
SDP
Catalog No. 501907305
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Fitting Type 23. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A23- DC Nebulizers. Compatible with PerkinElmer NexION 1000/2000/5000 ICP-MS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7305 Supplier Glass Expansion Supplier No. 708031449
Direct Connect (DC) Nebulizer Gas Fitting Type 23. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A23- DC Nebulizers. Compatible with PerkinElmer NexION 1000/2000/5000 ICP-MS.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001